Assay Details
| Gene Symbol: CYC1BAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342857.1 | NP_001332193.1 | 2 - 3 | 916 | 146 | |
| NM_120697.3 | NP_196233.1 | 2 - 3 | 963 | 146 | ||
More Information
Set Membership: |
|

CYC1BAT
Genomic Map