Assay Details
| Gene Symbol: DPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125705.2 | NP_001119177.1 | 1 - 2 | 680 | 69 | |
| NM_001342863.1 | NP_001331852.1 | 1 - 2 | 909 | 69 | ||
| NM_001342864.1 | NP_001331851.1 | 1 - 2 | 909 | 69 | ||
More Information
Set Membership: |
|

DPA4
Genomic Map