Assay Details
| Gene Symbol: 2-Cys Prx B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342875.1 | NP_001331225.1 | 4 - 5 | 664 | 100 | |
| NM_120712.4 | NP_568166.1 | 4 - 5 | 763 | 100 | ||
| GenBank mRNA | AF326871.1 | - | 4 - 5 | 470 | 100 | |
| AF339693.1 | - | 4 - 5 | 463 | 100 | ||
| AY054621.1 | - | 4 - 5 | 595 | 100 | ||
| AY081503.1 | - | 4 - 5 | 463 | 100 | ||
More Information
Set Membership: |
|

2-Cys Prx B
Genomic Map