Assay Details
Gene Symbol: 2-Cys Prx B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342875.1 | NP_001331225.1 | 5 - 6 | 791 | 115 | |
NM_120712.4 | NP_568166.1 | 5 - 6 | 890 | 115 | ||
GenBank mRNA | AF326871.1 | - | 5 - 6 | 597 | 115 | |
AF339693.1 | - | 5 - 6 | 590 | 115 | ||
AY054621.1 | - | 5 - 6 | 722 | 115 | ||
AY081503.1 | - | 5 - 6 | 590 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
