Assay Details
| Gene Symbol: UBA 2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120729.4 | NP_568168.1 | 5 - 6 | 2331 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBA 2| Gene Symbol: UBA 2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120729.4 | NP_568168.1 | 5 - 6 | 2331 | 84 | |
Set Membership: |
|