Assay Details
| Gene Symbol: AGL96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120733.2 | NP_196268.1 | 1 | 696 | 89 | |
| GenBank mRNA | DQ056672.1 | - | 1 | 696 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL96| Gene Symbol: AGL96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120733.2 | NP_196268.1 | 1 | 696 | 89 | |
| GenBank mRNA | DQ056672.1 | - | 1 | 696 | 89 | |
Set Membership: |
|