Assay Details
| Gene Symbol: NF-YA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342881.1 | NP_001330575.1 | 3 - 4 | 591 | 90 | |
| NM_001342882.1 | NP_001330576.1 | 3 - 4 | 579 | 90 | ||
| NM_120734.4 | NP_196269.1 | 3 - 4 | 557 | 90 | ||
| NM_203012.1 | NP_974741.1 | 2 - 3 | 627 | 90 | ||
| NM_203013.3 | NP_974742.1 | 2 - 3 | 643 | 90 | ||
More Information
Set Membership: |
|

NF-YA10
Genomic Map