Assay Details
| Gene Symbol: NF-YA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342880.1 | NP_001330574.1 | 2 - 3 | 1026 | 69 | |
| NM_001342881.1 | NP_001330575.1 | 4 - 5 | 793 | 69 | ||
| NM_001342882.1 | NP_001330576.1 | 4 - 5 | 781 | 69 | ||
| NM_001342883.1 | NP_001330577.1 | 1 - 2 | 259 | 69 | ||
| NM_120734.4 | NP_196269.1 | 4 - 5 | 759 | 69 | ||
| NM_203012.1 | NP_974741.1 | 3 - 4 | 829 | 69 | ||
| NM_203013.3 | NP_974742.1 | 3 - 4 | 845 | 69 | ||
More Information
Set Membership: |
|

NF-YA10
Genomic Map