Assay Details
| Gene Symbol: SPC98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120751.4 | NP_196286.1 | 1 - 2 | 2463 | 95 | |
| GenBank mRNA | AY099727.1 | - | 1 - 2 | 2404 | 95 | |
| BT010545.1 | - | 1 - 2 | 2250 | 95 | ||
More Information
Set Membership: |
|

SPC98
Genomic Map