Assay Details
| Gene Symbol: SRF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120765.3 | NP_196300.1 | 4 - 5 | 638 | 83 | |
| GenBank mRNA | AY518287.1 | - | 4 - 5 | 485 | 83 | |
| FJ708770.1 | - | 4 - 5 | 360 | 83 | ||
More Information
Set Membership: |
|

SRF2
Genomic Map