Assay Details
| Gene Symbol: AT5G07230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120805.4 | NP_196340.1 | 1 - 2 | 330 | 75 | |
| GenBank mRNA | AK175994.1 | - | 1 - 2 | 313 | 75 | |
More Information
Set Membership: |
|

AT5G07230
Genomic Map