Assay Details
| Gene Symbol: APC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120814.3 | NP_196349.1 | 3 - 4 | 1652 | 74 | |
| GenBank mRNA | AK175228.1 | - | 3 - 4 | 1604 | 74 | |
More Information
Set Membership: |
|

APC3
Genomic Map