Assay Details
| Gene Symbol: Tudor1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161225.2 | NP_001154697.2 | 13 - 14 | 2283 | 113 | |
| NM_120817.3 | NP_196352.2 | 13 - 14 | 2399 | 113 | ||
| GenBank mRNA | AY064975.1 | - | 13 - 14 | 2283 | 113 | |
More Information
Set Membership: |
|

Tudor1
Genomic Map