Assay Details
| Gene Symbol: GDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125711.2 | NP_001119183.1 | 5 - 6 | 916 | 66 | |
| NM_001125712.2 | NP_001119184.1 | 4 - 5 | 824 | 66 | ||
| NM_120826.3 | NP_196361.1 | 4 - 5 | 1027 | 66 | ||
More Information
Set Membership: |
|

GDH2
Genomic Map