Assay Details
| Gene Symbol: GDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125711.2 | NP_001119183.1 | 7 - 8 | 1243 | 85 | |
| NM_120826.3 | NP_196361.1 | 6 - 7 | 1354 | 85 | ||
| GenBank mRNA | U56635.1 | - | 6 - 7 | 1178 | 85 | |
More Information
Set Membership: |
|

GDH2
Genomic Map