Assay Details
| Gene Symbol: KUOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342944.1 | NP_001318498.1 | 3 - 4 | 796 | 141 | |
| NM_001342945.1 | NP_001332701.1 | 3 - 4 | 594 | 141 | ||
More Information
Set Membership: |
|

KUOX1
Genomic Map