Assay Details
| Gene Symbol: GRP18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120834.5 | NP_196369.1 | 1 - 2 | 331 | 104 | |
| GenBank mRNA | DQ446929.1 | - | 1 - 2 | 244 | 104 | |
More Information
Set Membership: |
|

GRP18
Genomic Map