Assay Details
| Gene Symbol: GRP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120838.4 | NP_196373.1 | 1 - 2 | 704 | 81 | |
| GenBank mRNA | BT004110.1 | - | 1 - 2 | 334 | 81 | |
| BT005466.1 | - | 1 - 2 | 309 | 81 | ||
More Information
Set Membership: |
|

GRP20
Genomic Map