Assay Details
| Gene Symbol: FH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120860.3 | NP_196395.1 | 12 - 13 | 1413 | 71 | |
| GenBank mRNA | AK221526.1 | - | 12 - 13 | 1413 | 71 | |
More Information
Set Membership: |
|

FH19
Genomic Map