Assay Details
| Gene Symbol: FH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120860.3 | NP_196395.1 | 5 - 6 | 831 | 74 | |
| GenBank mRNA | AK221526.1 | - | 5 - 6 | 831 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FH19| Gene Symbol: FH19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120860.3 | NP_196395.1 | 5 - 6 | 831 | 74 | |
| GenBank mRNA | AK221526.1 | - | 5 - 6 | 831 | 74 | |
Set Membership: |
|