Assay Details
| Gene Symbol: DGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342967.1 | NP_001331084.1 | 6 - 7 | 2427 | 74 | |
| NM_001342968.1 | NP_001331085.1 | 6 - 7 | 2239 | 74 | ||
| NM_120874.3 | NP_196409.1 | 6 - 7 | 2427 | 74 | ||
| GenBank mRNA | BT004148.1 | - | 6 - 7 | 2336 | 74 | |
More Information
Set Membership: |
|

DGK1
Genomic Map