Assay Details
| Gene Symbol: DGK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342968.1 | NP_001331085.1 | 7 - 8 | 2375 | 68 | |
| NM_120874.3 | NP_196409.1 | 7 - 8 | 2563 | 68 | ||
| GenBank mRNA | BT004148.1 | - | 7 - 8 | 2472 | 68 | |
More Information
Set Membership: |
|

DGK1
Genomic Map