Assay Details
| Gene Symbol: TT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120881.3 | NP_196416.1 | 1 - 2 | 556 | 107 | |
| GenBank mRNA | AF271650.1 | - | 1 - 2 | 447 | 107 | |
| AF271651.1 | - | 1 - 2 | 447 | 107 | ||
More Information
Set Membership: |
|

TT7
Genomic Map