Assay Details
| Gene Symbol: GDPD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342976.1 | NP_001331075.1 | 4 - 5 | 613 | 141 | |
| NM_120885.4 | NP_196420.1 | 4 - 5 | 608 | 141 | ||
| GenBank mRNA | BT023450.1 | - | 4 - 5 | 575 | 141 | |
More Information
Set Membership: |
|

GDPD6
Genomic Map