Assay Details
| Gene Symbol: TOM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120886.3 | NP_196421.1 | 1 - 2 | 323 | 79 | |
| GenBank mRNA | BT010778.1 | - | 1 - 2 | 132 | 79 | |
| BT011252.1 | - | 1 - 2 | 86 | 79 | ||
More Information
Set Membership: |
|

TOM5
Genomic Map