Assay Details
| Gene Symbol: SYP132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203327.1 | NP_001190256.1 | 11 - 12 | 814 | 75 | |
| NM_001342979.1 | NP_001332203.1 | 11 - 12 | 823 | 75 | ||
| NM_120890.3 | NP_568187.1 | 10 - 11 | 963 | 75 | ||
| GenBank mRNA | AY063836.1 | - | 10 - 11 | 777 | 75 | |
| AY117244.1 | - | 10 - 11 | 591 | 75 | ||
More Information
Set Membership: |
|

SYP132
Genomic Map