Assay Details
| Gene Symbol: SYP132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036776.1 | NP_001031853.1 | 3 - 4 | 366 | 132 | |
| NM_001342978.1 | NP_001318506.1 | 4 - 5 | 405 | 132 | ||
| NM_001342979.1 | NP_001332203.1 | 4 - 5 | 408 | 132 | ||
| NM_120890.3 | NP_568187.1 | 3 - 4 | 548 | 132 | ||
| GenBank mRNA | AY063836.1 | - | 3 - 4 | 362 | 132 | |
| AY117244.1 | - | 3 - 4 | 176 | 132 | ||
More Information
Set Membership: |
|

SYP132
Genomic Map