Assay Details
| Gene Symbol: EMB1873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342988.1 | NP_001330496.1 | 5 - 6 | 686 | 104 | |
| NM_001342989.1 | NP_001330497.1 | 4 - 5 | 766 | 104 | ||
| NM_001342990.1 | NP_001330494.1 | 5 - 6 | 666 | 104 | ||
| NM_001342991.1 | NP_001330495.1 | 5 - 6 | 519 | 104 | ||
| NM_120900.6 | NP_196434.1 | 5 - 6 | 686 | 104 | ||
| GenBank mRNA | AX765856.1 | - | 5 - 6 | 438 | 104 | |
More Information
Set Membership: |
|

EMB1873
Genomic Map