Assay Details
| Gene Symbol: EMB1873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342988.1 | NP_001330496.1 | 8 - 9 | 1126 | 76 | |
| NM_001342989.1 | NP_001330497.1 | 7 - 8 | 1206 | 76 | ||
| NM_001342990.1 | NP_001330494.1 | 8 - 9 | 1106 | 76 | ||
| NM_001342991.1 | NP_001330495.1 | 8 - 9 | 959 | 76 | ||
| NM_120900.6 | NP_196434.1 | 8 - 9 | 1126 | 76 | ||
| GenBank mRNA | AX765856.1 | - | 8 - 9 | 878 | 76 | |
More Information
Set Membership: |
|

EMB1873
Genomic Map