Assay Details
| Gene Symbol: AT5G08230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120906.6 | NP_196440.1 | 3 - 4 | 2456 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G08230| Gene Symbol: AT5G08230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120906.6 | NP_196440.1 | 3 - 4 | 2456 | 111 | |
Set Membership: |
|