Assay Details
| Gene Symbol: SULA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335740.1 | NP_001324767.1 | 9 - 10 | 763 | 79 | |
| NM_127700.4 | NP_565505.1 | 9 - 10 | 694 | 79 | ||
| GenBank mRNA | AF324701.2 | - | 9 - 10 | 681 | 79 | |
| AF326895.1 | - | 9 - 10 | 681 | 79 | ||
| AF339713.1 | - | 9 - 10 | 666 | 79 | ||
| AJ516948.1 | - | 9 - 10 | 666 | 79 | ||
| BT000757.1 | - | 9 - 10 | 681 | 79 | ||
More Information
Set Membership: |
|

SULA
Genomic Map