Assay Details
| Gene Symbol: FBA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036309.2 | NP_001031386.1 | 1 - 2 | 268 | 110 | |
| NM_001036310.1 | NP_001031387.1 | 1 - 2 | 240 | 110 | ||
| NM_127705.4 | NP_565508.1 | 1 - 2 | 647 | 110 | ||
| GenBank mRNA | AF419589.1 | - | 1 - 2 | 236 | 110 | |
| AF428408.1 | - | 1 - 2 | 236 | 110 | ||
| AY049282.1 | - | 1 - 2 | 236 | 110 | ||
| AY049286.1 | - | 1 - 2 | 236 | 110 | ||
| AY062582.1 | - | 1 - 2 | 236 | 110 | ||
| AY090291.1 | - | 1 - 2 | 180 | 110 | ||
| AY128380.1 | - | 1 - 2 | 238 | 110 | ||
| AY128784.1 | - | 1 - 2 | 180 | 110 | ||
| AY142633.1 | - | 1 - 2 | 180 | 110 | ||
| BT000106.1 | - | 1 - 2 | 180 | 110 | ||
| BT002415.1 | - | 1 - 2 | 236 | 110 | ||
More Information
Set Membership: |
|

FBA1
Genomic Map