Assay Details
| Gene Symbol: XK-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335744.1 | NP_001323495.1 | 7 - 8 | 1240 | 78 | |
| NM_001335745.1 | NP_001323494.1 | 7 - 8 | 1253 | 78 | ||
| NM_127708.3 | NP_179732.2 | 7 - 8 | 1253 | 78 | ||
| NM_201776.3 | NP_973505.1 | 7 - 8 | 1240 | 78 | ||
| GenBank mRNA | AK175657.1 | - | 7 - 8 | 1174 | 78 | |
| AK176783.1 | - | 7 - 8 | 1174 | 78 | ||
| AY136397.1 | - | 7 - 8 | 1176 | 78 | ||
| BT000210.1 | - | 7 - 8 | 1161 | 78 | ||
More Information
Set Membership: |
|

XK-1
Genomic Map