Assay Details
| Gene Symbol: VHA-A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127713.4 | NP_179736.1 | 1 - 2 | 342 | 76 | |
| GenBank mRNA | AF424556.1 | - | 1 - 2 | 191 | 76 | |
| AY099690.1 | - | 1 - 2 | 234 | 76 | ||
More Information
Set Membership: |
|

VHA-A2
Genomic Map