Assay Details
| Gene Symbol: APL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036314.2 | NP_001031391.1 | 12 - 13 | 1833 | 71 | |
| NM_001335765.1 | NP_001324982.1 | 12 - 13 | 1616 | 71 | ||
| NM_001335766.1 | NP_001324978.1 | 11 - 12 | 1934 | 71 | ||
| NM_001335767.1 | NP_001324979.1 | 11 - 12 | 1597 | 71 | ||
| NM_001335768.1 | NP_001324980.1 | 11 - 12 | 1284 | 71 | ||
| NM_001335769.1 | NP_001324981.1 | 10 - 11 | 1251 | 71 | ||
| NM_127730.5 | NP_179753.1 | 11 - 12 | 1550 | 71 | ||
| GenBank mRNA | AK317139.1 | - | 12 - 13 | 1452 | 71 | |
| AY070429.1 | - | 11 - 12 | 1318 | 71 | ||
| AY096657.1 | - | 10 - 11 | 1192 | 71 | ||
More Information
Set Membership: |
|

APL4
Genomic Map