Assay Details
| Gene Symbol: APL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036314.2 | NP_001031391.1 | 3 - 4 | 866 | 77 | |
| NM_001335765.1 | NP_001324982.1 | 3 - 4 | 649 | 77 | ||
| NM_001335766.1 | NP_001324978.1 | 2 - 3 | 967 | 77 | ||
| NM_001335767.1 | NP_001324979.1 | 2 - 3 | 630 | 77 | ||
| NM_001335768.1 | NP_001324980.1 | 2 - 3 | 317 | 77 | ||
| NM_001335769.1 | NP_001324981.1 | 1 - 2 | 284 | 77 | ||
| NM_127730.5 | NP_179753.1 | 2 - 3 | 583 | 77 | ||
| GenBank mRNA | AK317139.1 | - | 3 - 4 | 485 | 77 | |
| AY070429.1 | - | 2 - 3 | 351 | 77 | ||
| AY096657.1 | - | 1 - 2 | 225 | 77 | ||
More Information
Set Membership: |
|

APL4
Genomic Map