Assay Details
| Gene Symbol: APL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036314.2 | NP_001031391.1 | 7 - 8 | 1330 | 126 | |
| NM_001335765.1 | NP_001324982.1 | 7 - 8 | 1113 | 126 | ||
| NM_001335766.1 | NP_001324978.1 | 6 - 7 | 1431 | 126 | ||
| NM_001335767.1 | NP_001324979.1 | 6 - 7 | 1094 | 126 | ||
| NM_001335768.1 | NP_001324980.1 | 6 - 7 | 781 | 126 | ||
| NM_001335769.1 | NP_001324981.1 | 5 - 6 | 748 | 126 | ||
| NM_127730.5 | NP_179753.1 | 6 - 7 | 1047 | 126 | ||
| GenBank mRNA | AK317139.1 | - | 7 - 8 | 949 | 126 | |
| AY070429.1 | - | 6 - 7 | 815 | 126 | ||
| AY096657.1 | - | 5 - 6 | 689 | 126 | ||
More Information
Set Membership: |
|

APL4
Genomic Map