Assay Details
| Gene Symbol: AT2G21630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127734.3 | NP_179757.1 | 5 - 6 | 1518 | 77 | |
| GenBank mRNA | AY099755.1 | - | 5 - 6 | 1495 | 77 | |
| BT010345.1 | - | 5 - 6 | 1470 | 77 | ||
More Information
Set Membership: |
|

AT2G21630
Genomic Map