Assay Details
| Gene Symbol: RNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127748.4 | NP_179770.1 | 10 - 11 | 1723 | 146 | |
| GenBank mRNA | AY035080.1 | - | 10 - 11 | 1629 | 146 | |
| BT008573.1 | - | 10 - 11 | 1455 | 146 | ||
More Information
Set Membership: |
|

RNR1
Genomic Map