Assay Details
| Gene Symbol: RNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127748.4 | NP_179770.1 | 13 - 14 | 2214 | 87 | |
| GenBank mRNA | AY035080.1 | - | 13 - 14 | 2120 | 87 | |
| BT008573.1 | - | 13 - 14 | 1946 | 87 | ||
More Information
Set Membership: |
|

RNR1
Genomic Map