Assay Details
| Gene Symbol: RNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127748.4 | NP_179770.1 | 6 - 7 | 1234 | 71 | |
| GenBank mRNA | AY035080.1 | - | 6 - 7 | 1140 | 71 | |
| BT008573.1 | - | 6 - 7 | 966 | 71 | ||
More Information
Set Membership: |
|

RNR1
Genomic Map