Assay Details
| Gene Symbol: MGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127756.4 | NP_179778.1 | 2 - 3 | 387 | 79 | |
| NM_179687.1 | NP_850018.1 | 2 - 3 | 254 | 79 | ||
| GenBank mRNA | AY046020.1 | - | 2 - 3 | 202 | 79 | |
| AY079312.1 | - | 2 - 3 | 138 | 79 | ||
More Information
Set Membership: |
|

MGP1
Genomic Map