Assay Details
| Gene Symbol: MGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127756.4 | NP_179778.1 | 3 - 4 | 526 | 82 | |
| NM_179687.1 | NP_850018.1 | 3 - 4 | 393 | 82 | ||
| GenBank mRNA | AY046020.1 | - | 3 - 4 | 341 | 82 | |
| AY079312.1 | - | 3 - 4 | 277 | 82 | ||
More Information
Set Membership: |
|

MGP1
Genomic Map