Assay Details
| Gene Symbol: MGP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127756.4 | NP_179778.1 | 5 - 6 | 785 | 90 | |
| NM_179687.1 | NP_850018.1 | 5 - 6 | 652 | 90 | ||
| GenBank mRNA | AY046020.1 | - | 5 - 6 | 600 | 90 | |
| AY079312.1 | - | 5 - 6 | 536 | 90 | ||
More Information
Set Membership: |
|

MGP1
Genomic Map