Assay Details
| Gene Symbol: SK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084467.2 | NP_001077936.1 | 6 - 7 | 833 | 108 | |
| NM_001084468.1 | NP_001077937.1 | 6 - 7 | 768 | 108 | ||
| NM_001084469.1 | NP_001077938.1 | 5 - 6 | 594 | 108 | ||
| NM_001335780.1 | NP_001323559.1 | 6 - 7 | 586 | 108 | ||
| NM_001335781.1 | NP_001323558.1 | 5 - 6 | 631 | 108 | ||
| NM_127763.4 | NP_179785.2 | 6 - 7 | 600 | 108 | ||
| NM_201778.3 | NP_973507.2 | 6 - 7 | 833 | 108 | ||
More Information
Set Membership: |
|

SK1
Genomic Map