Assay Details
| Gene Symbol: SK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084467.2 | NP_001077936.1 | 7 - 8 | 910 | 74 | |
| NM_001084468.1 | NP_001077937.1 | 7 - 8 | 845 | 74 | ||
| NM_001084469.1 | NP_001077938.1 | 6 - 7 | 671 | 74 | ||
| NM_001335780.1 | NP_001323559.1 | 7 - 8 | 663 | 74 | ||
| NM_001335781.1 | NP_001323558.1 | 6 - 7 | 708 | 74 | ||
| NM_127763.4 | NP_179785.2 | 7 - 8 | 677 | 74 | ||
| NM_201778.3 | NP_973507.2 | 7 - 8 | 910 | 74 | ||
More Information
Set Membership: |
|

SK1
Genomic Map