Assay Details
| Gene Symbol: SK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084468.1 | NP_001077937.1 | 9 - 10 | 1062 | 77 | |
| NM_001084469.1 | NP_001077938.1 | 8 - 9 | 888 | 77 | ||
| NM_127763.4 | NP_179785.2 | 9 - 10 | 894 | 77 | ||
| NM_201778.3 | NP_973507.2 | 9 - 10 | 1127 | 77 | ||
More Information
Set Membership: |
|

SK1
Genomic Map