Assay Details
| Gene Symbol: AAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036317.1 | NP_001031394.1 | 8 - 9 | 1110 | 71 | |
| NM_127791.3 | NP_565529.1 | 7 - 8 | 1613 | 71 | ||
| NM_179691.4 | NP_850022.1 | 8 - 9 | 1046 | 71 | ||
| GenBank mRNA | AY064152.1 | - | 7 - 8 | 1095 | 71 | |
| AY124811.1 | - | 7 - 8 | 1002 | 71 | ||
| HM638413.1 | - | 7 - 8 | 1002 | 71 | ||
More Information
Set Membership: |
|

AAT
Genomic Map