Assay Details
| Gene Symbol: SLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127805.3 | NP_179827.1 | 1 | 2136 | 116 | |
| GenBank mRNA | AY064984.1 | - | 1 | 2135 | 116 | |
| HQ244435.1 | - | 1 | 2126 | 116 | ||
More Information
Set Membership: |
|

SLO1
Genomic Map