Assay Details
| Gene Symbol: CYCD2;1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202647.1 | NP_001189576.1 | 3 - 4 | 770 | 144 | |
| NM_127815.4 | NP_179835.1 | 3 - 4 | 792 | 144 | ||
| GenBank mRNA | BT025242.1 | - | 3 - 4 | 498 | 144 | |
| X83370.1 | - | 692 | 144 | |||
More Information
Set Membership: |
|

CYCD2;1
Genomic Map